Fluke TP88 Rigid Back Probe Pins
Representative image only.
Fluke TP88 Rigid Back Probe Pins
Fluke | Mfr: TP88| Item: 2003633
Helps take precise readings at difficult-to-measure locations
$67.78CAD / Each
$67.78CAD / Each
Product Description
Key Features
  • Slide on test probes measuring 2 mm
  • 2" long with sharp point—easily pass between the weather pack seal and wire
  • Insulation protects pins from shorting to other wires
  • Tested to 60 V DC
  • 1 year warranty
More About this Item
Slide on test probes measuring 2 mm to extend probe length to take measurements in difficult locations. Probe pins feature a sharp point to ensure a good contact to the electrical connector.
1 Year Warranty
Specifications
  • Compatible With2 mm probes
  • DescriptionRigid Back Probe Pins