Fluke TL71 Premium Digital Multimeter Right Angle Test Lead Set
Representative image only.
Fluke TL71 Premium Digital Multimeter Right Angle Test Lead Set
Fluke | Mfr: TL71| Item: UZ-20008-50
High performance at an economical price
$67.78CAD / Each
$67.78CAD / Each
Buy It Together
Current Item
Fluke TL71 Premium Digital Multimeter Right Angle Test Lead Set
Fluke TL71 Premium Digital Multimeter Right Angle Test Lead Set
2000850
$67.78CAD / Each
+ =
Total Price 428.76 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Flexible silicone insulated leads are heat and cold resistant
  • Distinctive comfort grip probes
  • Recommended for microvolt measurements
  • CAT II 1000 V, 10 A rated without cap; CAT IV 600 V, CAT III 1000 V with protective cap
  • 1 year warranty
More About this Item
Various test leads are available to suit your application.
Comfort grip probes with right-angle test leads have WearGuard™ indicator for damage detection.
Available for sale in the United States and Canada only.
1 Year WarrantyCanadian Standards Association
Specifications
  • Safety RatingCAT II 1000 V, 10 A rated without cap; CAT IV 600 V, CAT III 1000 V with protective cap
  • DescriptionPremium Digital Multimeter Right Angle Test Lead Set