Fluke T5-1K/PRV240 Proving Unit Kit with T5-1000 Electrical Tester
Representative image only.
Fluke T5-1K/PRV240 Proving Unit Kit with T5-1000 Electrical Tester
Fluke | Mfr: FLUKE-T5-1K/PRV240| Item: 2004955
Validate the functionality of test tools without placing yourself in a potentially hazardous electrical environment
$591.85CAD / Each
$591.85CAD / Each
Product Description
Includes
See individual model for included contents.
Key Features
  • “Test before touch”—reduce unnecessary exposure to shock and arc flash hazards
  • Compatible with high or low impedance multimeters, clamp meters, or two pole testers
  • Sources both AC and DC steady-state voltage
  • 3 year warranty
More About this Item
Fluke proving unit kits combine the PRV420 proving unit with either a Fluke multimeter, clamp meter, or electrical tester providing you optimal flexibility based on your application requirements. These kits are ideal for electrical maintenance personnel, electrical contractors, and electricians in the electrical, oil and gas, utilities markets, and more.
These kits, with included PRV240 proving unit, are designed for safety and compliance applying the concept of test before touch (TBT) that involves testing your meter against a known live source before and after measurements. This verifies that your test tool is operating properly during the actual measurement without exposing yourself to shock or arc flash.
The FLUKE-T5-1K/PRV240 includes PRV240 proving unit, T5-1000 tester, Probe tips, magnetic hanger strap, and two AA batteries.
For Sale in US and Canada Only
3 Year Warranty
Specifications
  • DescriptionProving Unit Kit with T5-1000 Electrical Tester
  • Warranty3 Years