Fluke T+PRO Electrical Voltage Tester with Backlit Display
Representative image only.
Fluke T+PRO Electrical Voltage Tester with Backlit Display
Fluke | Mfr: T+PRO| Item: 2604894
Call For Price
for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Fluke T+PRO Electrical Voltage Tester with Backlit Display
Fluke T+PRO Electrical Voltage Tester with Backlit Display
2604894
Call For Price
+ =
Total Price 195.59 ADD 2 ITEMS TO CART
Product Description
More About this Item
The Fluke T + PRO is the ideal, full-featured tester for the demanding commercial and industrial electrician. The Fluke T + is the basic electrical tester for residential electricians and HVAC technicians. These testers include all of the advantages of a traditional solenoid tester, with added functionality, but none of the typical drawbacks. With voltage measurement, continuity, built-in flashlights, GFCI trip capability, and Fluke's legendary ruggedness and reliability, the tester is an excellent choice for most residential, commercial, and industrial applications.
  • Safer than traditional solenoid testers
    Available for sale in the United States and Canada only.
  • Specifications
    • Safety RatingCAT III 1000V, CAT IV 600V, IEC 1010-1
    • DescriptionElectrical Voltage Tester with Backlit Display