Fluke FLK-A3002 FC Wireless AC/DC Current Module

Representative image only.

Fluke FLK-A3002 FC Wireless AC/DC Current Module

Fluke | Mfr: FLK-A3002 FC| Item: 2004780

See, save, and share data wirelessly — saving time and increasing productivity

$284.35CAD / Each

for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$284.35CAD / Each

Buy It Together

Current Item

Fluke FLK-A3002 FC Wireless AC/DC Current Module
Fluke FLK-A3002 FC Wireless AC/DC Current Module
2004780

$284.35CAD / Each
+ =
Total Price 479.94 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty

More About this Item

The Fluke A3002 FC module measures up to 600 A AC or 1000 A DC and can be used as a standalone meter or as part of the Fluke connect wireless system. The logging function records and saves up to 65,000 readings for added flexibility.

The Fluke A3002 FC AC/DC current module wirelessly relays measurements from clamp accessory to other Fluke Connect enabled master units listed below:
Fluke 3000 FC DMM
Fluke Ti200/300/400Infrared Cameras
To your PC via optional PC3000 FC Adapter

What’s included: Magnetic hanging strap and two AA batteries.

Available for sale in the United States and Canada only

3 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • DC Current Accuracy0.5% ±3 digits
  • Current (AC/DC)600 A AC / 1000 A DC
  • AC Current Accuracy1.0% ±3 digits
  • Width (in)2.33
  • Height (in)6.33
  • Depth (in)1.5
  • Width (cm)6.6
  • Height (cm)16
  • Depth (cm)3.8
  • DescriptionWireless AC/DC Current Module
  • Warranty3 Years
Reviews
Q&A