Fluke FLK-A3001 FC FC Wireless iFlex AC Current Module

Representative image only.

Fluke FLK-A3001 FC FC Wireless iFlex AC Current Module

Fluke | Mfr: FLK-A3001 FC| Item: 2004776

See, save, and share data wirelessly — saving time and increasing productivity

$545.14CAD / Each

for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$545.14CAD / Each

Buy It Together

Current Item

Fluke FLK-A3001 FC FC Wireless iFlex AC Current Module
Fluke FLK-A3001 FC FC Wireless iFlex AC Current Module
2004776

$545.14CAD / Each
+ =
Total Price 740.73 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty

More About this Item

The Fluke A3001 FC flexible wireless current module allows you to measure around awkward sized conductors and get into tight spaces for easier access to wires. Use as a standalone meter or as part of the Fluke connect wireless system
The logging function allows recording and saving up to 65,000 readings and inrush function provides additional flexibility.

The Fluke A3001 FC True-rms current clamp meter wirelessly relays measurements to other Fluke Connect™ enabled master units listed below:
Fluke 3000 FC DMM
Fluke Ti200/300/400 Infrared Cameras
To your PC via optional PC3000 FC adapter

Whats included:
One true-rms flexible current meter and one iFlex i2500-10 flexible current probe, magnetic hanging strap, and two AA batteries.

Available for sale in the United States and Canada only

3 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • AC Max (amps)2500
  • AC Current Accuracy3% ±5 digits
  • Width (in)2.5
  • Height (in)6.5
  • Depth (in)1.5
  • Width (cm)6.35
  • Height (cm)16.5
  • Depth (cm)3.8
  • DescriptionFC Wireless iFlex AC Current Module
  • Warranty3 Years
Reviews
Q&A