Fluke FLK-A3000 FC Wireless AC Current Module

Representative image only.

Fluke FLK-A3000 FC Wireless AC Current Module

Fluke | Mfr: FLK-A3000 FC| Item: 2004777

See, save, and share data wirelessly — saving time and increasing productivity

$449.37CAD / Each

for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$449.37CAD / Each

Buy It Together

Current Item

Fluke FLK-A3000 FC Wireless AC Current Module
Fluke FLK-A3000 FC Wireless AC Current Module
2004777

$449.37CAD / Each
+ =
Total Price 644.96 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty

More About this Item

The Fluke A3000 FC True-rms ac clamp meter measures up to 400 A AC and can be used as a standalone meter or as part of the Fluke connect system. The data logging function allows you to record and save up to 65,000 readings and in rush function provides additional flexibility.

The Fluke A3000 FC is a fully functional true-rms current clamp meter that wirelessly relays measurements to other Fluke Connect enabled master units listed below:
Fluke 3000 FC DMM
Fluke Ti200/300/400 Infrared Cameras
To your PC via optional PC3000 FC Adapter

What’s included: Two AA batteries.

Available for sale in the United States and Canada only

3 Year Warranty
Specifications
  • AC Max (amps)400
  • AC Current Accuracy2% ±5 digits
  • Width (in)3
  • Height (in)8
  • Depth (in)1.5
  • Width (cm)7.49
  • Height (cm)20.3
  • Depth (cm)3.8
  • DescriptionWireless AC Current Module
  • Warranty3 Years
Reviews
Q&A