Fluke FLK-3000 FC Wireless Digital Multimeter

Representative image only.

Fluke FLK-3000 FC Wireless Digital Multimeter

Fluke | Mfr: FLK-3000 FC| Item: 2004775

See, save, and share data wirelessly — saving time and increasing productivity

$663.01CAD / Each

for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$663.01CAD / Each

Buy It Together

Current Item

Fluke FLK-3000 FC Wireless Digital Multimeter
Fluke FLK-3000 FC Wireless Digital Multimeter
2004775

$663.01CAD / Each
+ =
Total Price 858.60 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty

More About this Item


The new Fluke Connect system of test tools communicates with the new Fluke Connect mobile
app on your Android™ or iOS smart device, allowing you to share live data, monitor conditions from safe distances, and get your job done quicker and easier. The wireless digital multimeter functions as the mobile hub as it displays readings from up to three module tools simultaneously from up to 20 meters away. The system allows you to view up to six live readings at once with the Fluke connect mobile app or a PC, in addition to capturing up to 65,000 min/max/avg measurements wirelessly with the PC3000 FC adapter and software (order separately) , each time stamped, over a single or multiple logging sessions.
Select the required wireless digital multimeter and optional module(s) to complete your wireless system. The wireless enabled modules measure AC/DC current (choose a standard clamp or flexible clamp to measure AC current), temperature, and AC/DC voltage. Mix and match the modules, or select from preconfigured kits for additional cost savings and convenience based on your unique application requirements.

Whats included: 3000FC wireless digital multimeter includes TL175 test lead set, AC175 alligator clip set, magnetic hanger strap, and three AA batteries.

Available for sale in the United States and Canada only

3 Year WarrantyConformite EuropeenneIP54 Rated
Specifications
  • DC Min (Volts)0.0001
  • DC Max (Volts)1000
  • DC Voltage Accuracy0.09% ±2 digits
  • DC Min (amps)1E-05
  • DC Max (amps)0.4
  • DC Current Accuracy0.5% ±3 digits
  • AC Max (Volts)1000
  • Ac Voltage Accuracy1.0% ±3 digits
  • AC Max (amps)0.4
  • AC Current Accuracy1.5% ±3 digits
  • Resistance Max (Ω)600
  • Capacitance Max (nF)1000
  • Capacitance Min (mF)9999
  • Frequency Max (Hz)99.99
  • Continuity600 Ohms
  • PowerThree AA batteries
  • DescriptionWireless Digital Multimeter
  • Warranty3 Years
Reviews
Q&A