Fluke CO-220 Digital carbon monoxide meter
Representative image only.
Fluke CO-220 Digital carbon monoxide meter
Fluke | Mfr: FLUKE-CO-220| Item: 2602000
Comfortable ergonomic design and easy, one-button operation
$1,075.57CAD / Each
for an additional
$356.56CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,075.57CAD / Each
Buy It Together
Current Item
Fluke CO-220 Digital carbon monoxide meter
Fluke CO-220 Digital carbon monoxide meter
2602000
$1,075.57CAD / Each
+ =
Total Price 1432.13 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Rugged case and electronics
  • Bright backlight allows easy screen viewing under dim lighting conditions
  • Beeper triggers with increasing frequency as CO levels rise
  • MAX Hold function stores and displays the maximum CO level
  • Automatic sensor zeroing and self-test sequence upon start-up
  • CO level beeper can be silenced if desired
  • Automatic power-off after 20 minutes extends battery life
  • Easy to replace battery
  • Replaceable sensor extends tool life
  • 1 year warranty
More About this Item
Feature stabilized electrochemical CO-specific sensor that needs only annual recalibration. Choose from digital meter and indicator probe--both include soft carrying case and battery. Order optional aspirator kit to collect air samples for analysis with either instrument.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • Gases MeasuredCO
  • Min CO Range (ppm)0
  • Max CO Range (ppm)1000
  • CO Accuracy±3% of reading
  • CO Resolution (ppm)1
  • DescriptionDigital carbon monoxide meter