Fluke Calibration TQSoft Thermal Validation Software with IQ/OQ Documents
Representative image only.
Fluke Calibration TQSoft Thermal Validation Software with IQ/OQ Documents
Fluke Calibration | Mfr: TQSOFT-IQ/OQ| Item: 1611400
Compliance made easy
$6,239.81CAD / Each
$6,239.81CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This calibration and validation software simplifies test setup with step-by-step guides and dropdown lists of available options. Automatic and manual sensor calibrations for multichannel temperature measurement systems determine mulitple point offsets for each channel. The software assists with equipment setup, such as data logger channel configuration and temperature source communications. Validation test development options include 3-D chamber space diagrams with multicolor temperature gradients based on data from multiple sensors and real-time temperature displays. Once complete, test data can be played back in graph form showing the change in the temperature map over time.
Sample tests are included for practice and to use as templates in devising your own tests. A large library of reporting templates make report generation quick and easy. From cover pages to calibration certificates, the templates can be merged into a full report and saved or printed. All authorized changes, from equipment preparation and sensor calibration to final reports, are documented with full details of the changes and any actions performed providing comprehensive data security.
Choose TQSoft for pharmaceutical process analysis and validation, or TQAero for aerospace furnace validation.
For sale in US and Canada only
1 Year Warranty
Specifications
  • Operating System VersionWindows® XP, 7, 8
  • DescriptionThermal Validation Software with IQ/OQ Documents