Fluke Calibration 9935-M Multi Channel LogWare Software
Representative image only.
Fluke Calibration 9935-M Multi Channel LogWare Software
Fluke Calibration | Mfr: 9935-M| Item: 1610316
Offers accurate, expandable, and configurable readouts
$2,806.81CAD / Each
$2,806.81CAD / Each
Product Description
Key Features
  • Able to read SPRTs, RTDs, thermistors, and thermocouples
  • Can be configured with up to eight modules
  • Serves as a high-accuracy reference thermometer, with accuracy up to ±0.0013 °C
  • 1 year warranty
More About this Item
The 1560 Black Stack Thermometer Readout can be any kind of thermometer you want it to be, and it works in three distinctive ways. It’s a reference thermometer with a NIST traceable calibration; it’s an automated calibration system reading your reference probe and sensors you’re testing; or it’s a high-accuracy data acquisition system.
The system consists of up to eight different modules (sold separately) designed to do any type of thermometry you choose. Each module stacks behind the preceding one, and when you add a module, the Stack’s software automatically reconfigures itself to include all of the new functions supplied by that module. There’s no software to load, and nothing has to be calibrated. Each module stores its own calibration internally, so you can add or change modules without recalibrating the whole stack. Module calibration is digital and is performed manually through the base’s front panel or over the RS-232 link.
The LCD screen has multiple methods of displaying data, including a graphical strip chart recorder. The graphical capability of the Black Stack makes testing temperature stability easier than ever. Vertical scaling and graph resolution are automatic. Read data almost anyway you like—in ohms, millivolts, or temperature, according to your application and preference.
The multi channel logWare software is a powerful temperature data acquisition tool. It provides graphical and statistical analysis of each channel you’re measuring (up to 96 with the Black Stack).
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • DescriptionSoftware Channel Multi User