Fluke Calibration 9132 Infrared digital calibrator
Representative image only.
Fluke Calibration 9132 Infrared digital calibrator
Fluke Calibration | Mfr: 9132-156| Item: 9091110
$10,350.56CAD / Each
for an additional
$1,321.63CAD / Each
for an additional
$1,196.39CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$10,350.56CAD / Each
Buy It Together
Current Item
Fluke Calibration 9132 Infrared digital calibrator
Fluke Calibration 9132 Infrared digital calibrator
9091110
$10,350.56CAD / Each
+ + =
Total Price 11234.23 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Calibrate over entire range — up to 932°F (500°C)!
  • Large 2-¼" diameter blackbody target provides a wide field of view—compensates for optical variations in your thermometers.
  • Included RTD reference well lets you check calibrator for accuracy.
  • Small, compact design takes up little space on your benchtop.
  • 1 year warranty
More About this Item
Check and maintain the accuracy of all your in-line, handheld, or benchtop infrared thermometers with this high-accuracy calibrator. The isothermal target can be controlled in increments of 0.1° from 122 to 932°F (50 to 500°C) by using the front panel controls. Calibration is simple: Set the desired calibration temperature via the front-panel mounted controls, then point your IR thermometer at the target. Compare the readings on the calibrator with the reading on your thermometer and record the difference. For higher precision calibration, use the RTD reference well with an optional RTD thermometer to check the accuracy of the calibrator. For a complete selection of RTD thermometers.
Available for sale in the United States and Canada only.
Infrared calibrator 90911-10 shown with infrared thermometer 39750-00 sold separately.
Hart Scientific is now Fluke Calibration.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Max Temperature (° F)932
  • Min Temperature (° F)122
  • Power (VAC)115
  • DescriptionInfrared digital calibrator