Fluke Calibration 9009-B Industrial Dual-Block Calibrator
Representative image only.
Fluke Calibration Mfr # 9009-B – Item # RK-16101-67
  • 1 year warranty
$12,498.77CAD / Each
Time to recalibrate?
for an additional
$682.18CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
  • DescriptionIndustrial Dual-Block Calibrator
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • Temperatures from -15°C to 350°C in one unit
  • Two wells in each block for simultaneous comparison calibrations
  • Rugged, lightweight, water-resistant enclosure
  • Controlled by precision temperature controller
This Dual-Block Calibrator lets you calibrate at hot and cold temperatures at the same time. Double your productivity or cut your calibration time in half—either way you look at it, your in-field temperature calibrations just got easier.
The unit includes two independently controlled temperature blocks. The hot block provides temperatures from 35°C to 350°C, while the cold block covers the range -15·°C to 110°C. Both blocks are controlled by a precision temperature controller.
Available for sale in the United States and Canada only.