Fluke Calibration 5611A-11-L Bare Thermistor Probe
Representative image only.
Fluke Calibration 5611A-11-L Bare Thermistor Probe
Fluke Calibration | Mfr: 5611A-11L| Item: 3785233
$1,684.53CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,684.53CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
5611A Series – Silicone-coated secondary thermistor probes: With a diameter at the tip of just 1.5 mm ( 1/16”), the 5611A''s tip has the smallest diameter of any of our secondary reference thermistors and can fit nearly anywhere. Its faster response time, flexible sheath, and silicone coating make the 5611A great for use in many applications. However, applications involving silicone oil could damage the thermistor and should be avoided.
Available for sale in the United States and Canada only.
Hart Scientific is now Fluke Calibration.
1 Year Warranty
Specifications
  • Probe TypeThermistor
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature Accuracy±0.018°F (±0.01°C)
  • Connector TypeMini Spade Lug
  • DescriptionBare Thermistor Probe