Fluke Calibration 5610-9-P SS Sheathed Secondary Reference Thermistor Probe, 9" L
Representative image only.
Fluke Calibration 5610-9-P SS Sheathed Secondary Reference Thermistor Probe, 9" L
Fluke Calibration | Mfr: 5610-9-P| Item: 9320253
Small diameter and very small sensing element results in fast response time
$2,084.85CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$2,084.85CAD / Each
Product Description
Key Features
  • Excellent for measuring temperatures in air, liquid or soil
  • Rugged polished stainless steel sheath offers durability
  • Short-term accuracy to ±0.01°C; one-year drift < ±0.01°C
  • Bare wire ends
  • 1 year warranty
More About this Item
Durable stainless steel thermistor probe offers excellent accuracy and fast response time.
Hart Scientific is now Fluke Calibration.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible With1504, 1502, and 1529 reference thermometers
  • Length (InchShort)9
  • DescriptionSS Sheathed Secondary Reference Thermistor Probe, 9" L