Fluke Calibration 2373-LTC Adapter, Lemo to Universal Thermocouple Probe
Representative image only.
Fluke Calibration 2373-LTC Adapter, Lemo to Universal Thermocouple Probe
Fluke Calibration | Mfr: 2373-LTC| Item: UZ-93202-58
Use to connect 1523 or 1524 reference thermometer to thermocouple leads
$611.46CAD / Each
$611.46CAD / Each
Product Description
Key Features
  • Supports reference junction compensation (RJC) with its own internal precision thermistor
  • 1 year warranty
More About this Item
Keep your thermometers running at their best with these accessories.
1 Year Warranty
Specifications
  • Compatible WithFluke Calibration 1523 and 1524 thermometers
  • ConnectionsLemo to universal thermocouple probe
  • DescriptionAdapter, Lemo to Universal Thermocouple Probe