Fluke Calibration 1586-2586-KIT High-Capacity Module and Relay Card Kit
Representative image only.
Fluke Calibration 1586-2586-KIT High-Capacity Module and Relay Card Kit
Fluke Calibration | Mfr: 1586-2586-KIT| Item: 9320123
1586A Scanner is designed to add one extra High-Capacity or Multiplexer module, adding 20 extra channels
$1,284.06CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$1,284.06CAD / Each
Buy It Together
Current Item
Fluke Calibration 1586-2586-KIT High-Capacity Module and Relay Card Kit
Fluke Calibration 1586-2586-KIT High-Capacity Module and Relay Card Kit
9320123
$1,284.06CAD / Each
+ =
Total Price 2383.21 ADD 2 ITEMS TO CART
Product Description
Includes
HC-model includes; one module, and relay card. Multiplexer-model includes; one module, adapter, and interface cable
Key Features
  • Modules can be mixed or matched to the 1586A Scanner
  • User can add a High-Capacity or Multiplexer module to either a 1586A High-Capacity or Multiplexer Scanner
  • Twenty 2-wire or ten 3-, or 4-wire connections
  • Two dedicated dc current channels
  • Built-in cable strain relief to help secure connections
  • Clear top case with embedded channel numbers for easy wiring identification
  • Can be used with either the 1586A High Capacity or Multiplexer scanners
  • 1 year warranty
More About this Item
Universal input channels designed to measure temperature, resistance, dc voltage, and dc current for data acquisition applications on the factory floor where channel count and scan speed are important.
Hart Scientific is now Fluke Calibration.
1 Year Warranty
Specifications
  • DescriptionHigh-Capacity Module and Relay Card Kit