Fluke Calibration 1551-CASE Carrying Case, for 1551 / 1552
Representative image only.
Fluke Calibration 1551-CASE Carrying Case, for 1551 / 1552
Fluke Calibration | Mfr: 1551-CASE| Item: 9320211
The best and most accurate way to replace glass thermometers
$212.46CAD / Each
$212.46CAD / Each
Product Description
Includes
NIST traceable NVLAP accredited report of calibration supplied by the manufacture, user’s guide on CD-ROM and 3-AAA batteries
Key Features
  • Built-in probe with a stainless steel sheath ideal for rugged applications
  • Display temperature in Celsius or Fahrenheit
  • ±0.05°C accuracy over full range
  • Temperature trend indicator
  • 200 hour battery life
  • 1 year warranty
More About this Item
Whether you’re verifying the accuracy of a temperature sensor or performing a procedure for the pharmaceutical industry, the Fluke Stik thermometer from Hart Scientific gives you a safe, portable, and accurate solution for all of your temperature calibration applications. The probe and digital readout are fixed together, calibrated as a system, and accurate to ±0.1°F (±0.05°C) over the full temperature range with selectable resolution of 0.1, 0.01 or 0.001. The instrument comes with an easy-to-read temperature trend indicator that you configure to meet your application’s stability requirements. The indicator tells you if your temperature is trending up or down and lets you know when your temperature source is stable enough for an accurate measurement. The highly accurate RTD probe is hinged and swivels 90° allowing you to easily view your temperature measurement from the large backlit LCD. The Stik Thermometer operates on three AAA batteries and includes a low-battery indicator and auto-off function that shuts down the thermometer to prevent erroneous measurements due to low battery life.
Available for sale in the United States and Canada only.
Hart Scientific is now Fluke Calibration.
1 Year WarrantyConformite Europeenne
Specifications
  • DescriptionCarrying Case, for 1551 / 1552