Fluke AC285 Large Jaw Alligator Clips
Representative image only.
Fluke AC285 Large Jaw Alligator Clips
Fluke | Mfr: AC285| Item: 2600186
High performance at an economical price
$73.65CAD / Set of 1
$73.65CAD / Set of 1
Product Description
Key Features
  • 1 year warranty
More About this Item
Fully insulated bananajack alligator clips. Large 0.750" (20 mm) jaw opening. One red, one black. 250 V, 10 A maximum.
Available for sale in the United States and Canada only
1 Year Warranty
Specifications
  • DescriptionLarge Jaw Alligator Clips