Fluke AC280 Hook-Style Test Clips
Representative image only.
Fluke AC280 Hook-Style Test Clips
Fluke | Mfr: AC280| Item: 2600181
High performance at an economical price
$89.86CAD / Set of 1
$89.86CAD / Set of 1
Product Description
Key Features
  • 1 year warranty
More About this Item
Safety-designed, spring-loaded clips with hook grip. Hook size is 0.400" x 0.080" (10 mm x 2 mm).
One red, one black.
1000 V, 3 A maximum.
Available for sale in the United States and Canada only
1 Year Warranty
Specifications
  • DescriptionHook-Style Test Clips