Fluke 80TK Thermocouple Module, DMM to Thermometer Converter
Representative image only.
Fluke 80TK Thermocouple Module, DMM to Thermometer Converter
Fluke | Mfr: 80TK| Item: 2605263
Convert a DMM to a thermometer
$344.76CAD / Each
$344.76CAD / Each
Product Description
Key Features
  • For use with type-K thermocouples in low voltage applications (below 24 V AC, 60 V DC)
  • Measurement range: -50 to 1000°C (probe dependent)
  • 1 year warranty
More About this Item
No need to purchase a thermocouple thermometer—connect the thermocouple module to your digital multimeter to take temperature measurements.
1 Year Warranty
Specifications
  • Compatible WithFluke DMMs, oscilloscopes, and loop calibrators
  • DescriptionThermocouple Module, DMM to Thermometer Converter