Fluke 80BK-A Type K, Integrated DMM Temperature Probe
Representative image only.
Fluke 80BK-A Type K, Integrated DMM Temperature Probe
Fluke | Mfr: 80BK-A| Item: 2000773
Connect to Fluke oscilloscopes and DMMs to easily measure temperatures
$109.03CAD / Each
$109.03CAD / Each
Product Description
Key Features
  • Type K probes
  • Type K thermocouple with standard banana jack connection
  • One piece construction with measurement range of -40 to 260ºC
  • Compatible with DMMs with temperature measurement functions
  • 1 year warranty
More About this Item
Compatible Fluke meters include:
Fluke 175 True RMS DMM
Fluke 177 True RMS DMM
Fluke 179 True RMS DMM
Fluke 28 II Ex Intrinsically Safe True RMS DMM
Fluke 287 True RMS Electronics Logging Multimeter
Fluke 289 True RMS Industrial Logging Multimeter
1 Year Warranty
Specifications
  • Compatible WithFluke digital multimeters
  • ConnectionsStandard shrouded banana jack
  • DescriptionType K, Integrated DMM Temperature Probe