Fluke 80AK-A Adapter for Suregrip Thermocouple Probes
Representative image only.
Fluke 80AK-A Adapter for Suregrip Thermocouple Probes
Fluke | Mfr: 80AK-A| Item: 9927898
Offers more flexible strain relief for a long life
$79.56CAD / Each
$79.56CAD / Each
Product Description
Key Features
  • Adapts Type-K thermocouple mini-connector to dual shrouded banana plug inputs
  • Measurement range and accuracy: probe dependent
  • Suitable for low voltage applications (below 30 V AC, 60 V DC)
  • 1 year warranty
More About this Item
The innovative SureGrip™ design has a soft rubber handle combined with a ergonomic shape is so comfortable to hold that you’ll forget about the probe and focus on the measurement. All SureGrip probes have an improved, more flexible strain relief for a long life.
1 Year Warranty
Specifications
  • Probe TypeK
  • Connector TypeMini-connector to dual shrouded banana plug
  • DescriptionAdapter for Suregrip Thermocouple Probes