Fluke 726 Precision Multi-Process Calibrator
Representative image only.
Fluke 726 Precision Multi-Process Calibrator
Fluke | Mfr: FLUKE-726| Item: 1610040
$8,692.99CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$8,692.99CAD / Each
Buy It Together
Current Item
Fluke 726 Precision Multi-Process Calibrator
Fluke 726 Precision Multi-Process Calibrator
1610040
$8,692.99CAD / Each
+ +
Fluke TPAK ToolPak Magnetic Meter Hanging Kit
2000856
$89.86CAD / Each
=
Total Price 9199.08 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Measure volts. mA, RTDs, thermocouples, frequency, and resistance to test sensors and transmitters
  • Source / simulate volts, mA, thermocouples, RTDs, frequency, and pressure to calibrate transmitters
  • Source mA with simultaneous pressure measurement to conduct valve and I/P tests
  • Perform fast linearity tests with auto step and auto ramp features
  • Power transmitters during test using 24 V loop supply and simultaneous mA measurement
  • Store frequently used test setups for later use
  • Available for sale in the United States and Canada only.
  • 3 year warranty
3 Year Warranty
Specifications
  • DescriptionPrecision Multi-Process Calibrator
  • Warranty3 Years