Fluke 717 300G Pressure Calibrator, 300 psig
Representative image only.
Fluke 717 300G Pressure Calibrator, 300 psig
Fluke | Mfr: FLUKE-717 300G| Item: 6845083
$3,528.39CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$3,528.39CAD / Each
Buy It Together
Current Item
Fluke 717 300G Pressure Calibrator, 300 psig
Fluke 717 300G Pressure Calibrator, 300 psig
6845083
$3,528.39CAD / Each
+ =
Total Price 3884.95 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3 year warranty
More About this Item
Pressure source and milliamp measurement capability to calibrate and maintain almost any pressure device with one tool
Error % calculation for quicker pass / fail decision making in the field
Best in class mA accuracy (0.015% for better e asuremetn confidence and workload coverage
3 Year Warranty
Specifications
  • Operating Temperature Range−10 °C to 55 °C
  • Min Pressure-12 psi
  • Pressure Accuracy0.05% of range
  • DescriptionPressure Calibrator, 300 psig
  • Warranty3 Years