Fluke 717 100G Pressure Calibrator, 100 psig
Representative image only.
Fluke 717 100G Pressure Calibrator, 100 psig
Fluke | Mfr: FLUKE-717 100G| Item: 6845079
$3,529.90CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$3,529.90CAD / Each
Buy It Together
Current Item
Fluke 717 100G Pressure Calibrator, 100 psig
Fluke 717 100G Pressure Calibrator, 100 psig
6845079
$3,529.90CAD / Each
+ =
Total Price 4011.70 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3 year warranty
More About this Item
Pressure source and milliamp measurement capability to calibrate and maintain almost any pressure device with one tool
Error % calculation for quicker pass / fail decision making in the field
Best in class mA accuracy (0.015% for better e asuremetn confidence and workload coverage
3 Year Warranty
Specifications
  • Operating Temperature Range−10 °C to 55 °C
  • Min Pressure-12 psi
  • Pressure Accuracy0.05% of range
  • DescriptionPressure Calibrator, 100 psig
  • Warranty3 Years