Fluke 700PTP-1 Pneumatic Test Pump, -13 to 600 psi
Representative image only.
Fluke 700PTP-1 Pneumatic Test Pump, -13 to 600 psi
Fluke | Mfr: FLUKE-700PTP-1| Item: 6845075
Generate pressure up to 600 psi accurately
$1,738.59CAD / Each
$1,738.59CAD / Each
Product Description
Includes
A ¼" NPT male to ¼" BSP female, 1 meter of hose, two 1/8" NPT male quick connects, 1/8" NPT female to ¼" BSP female, and a Seal kit.
Key Features
  • 1/4" NPT female fitting for the reference gauge or pressure module
  • 1/4" NPT female fitting for the unit under test
  • 1 year warranty
More About this Item
The Fluke 700PTP-1 pneumatic test pump is capable of supplying pressures up to 600 psi. For use with 700 series pressure modules or test gauges. Can be directly connected to the reference port consolidating two separate pieces when performing pressure calibration and testing. Can also be used with 717 pressure calibrators. The vernier can be used as a stand-alone low pressure calibration source.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • Max Pressure600 psi
  • DescriptionPneumatic Test Pump, -13 to 600 psi