Fluke 700PMP Hand-Operated Pneumatic Pump, 150 psi
Representative image only.
Fluke 700PMP Hand-Operated Pneumatic Pump, 150 psi
Fluke | Mfr: FLUKE-700PMP| Item: 6845074
Hand held pump generates pressure up to 150 psi accurately
$1,086.58CAD / Each
$1,086.58CAD / Each
Product Description
Key Features
  • Pump incorporated needle valve for fine venting adjustment
  • 1 year warranty
More About this Item
The Fluke 700PMP hand-operated pressure pump provides pressures up to 150 psi. It is a tube type, cylindrical shaped hand pump with a "T" handle at the compressor end and a round knob at the volume adjust end. The pump incorporates a needle valve for venting and a volume adjust vernier for precision adjustment of pressure. Pressure connections are made through one 1/8" NPT internally threaded fitting.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • Max Pressure150 psi
  • ConnectionsOne 1/4" NPT(F) port
  • DescriptionHand-Operated Pneumatic Pump, 150 psi