Fluke 117/323 KIT Electrician's Multimeter/Clamp Meter Combo Kit
Representative image only.
Fluke 117/323 KIT Electrician's Multimeter/Clamp Meter Combo Kit
Fluke | Mfr: FLUKE-117/323 KIT| Item: 2004602
Kit saves you time and money
$695.43CAD / Each
for an additional
$195.59CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$695.43CAD / Each
Buy It Together
Current Item
Fluke 117/323 KIT Electrician's Multimeter/Clamp Meter Combo Kit
Fluke 117/323 KIT Electrician's Multimeter/Clamp Meter Combo Kit
2004602
$695.43CAD / Each
+ =
Total Price 891.02 ADD 2 ITEMS TO CART
Product Description
Includes
117 True RMS digital multimeter with noncontact voltage detection, TL75 hardpoint test lead set, 323 True RMS clamp meter, ToolPak magnetic meter hanger for hands-free operation, C115 deluxe soft carrying case, and one 9 V and two AA batteries.
Key Features
  • Troubleshoot motors, drives, and panels quickly and accurately
  • Preventive maintenance kit reduces costly repairs and downtime
  • 2 year warranty
More About this Item
This combo kit combines the Fluke 117 multimeter with a Fluke 323 clamp meter for productive and effective troubleshooting in one total solution. This kit is an essential part of any organizations preventive maintenance program.
Available for sale in the United States and Canada only
Two year warranty
Specifications
  • DescriptionElectrician's Multimeter/Clamp Meter Combo Kit
  • Warranty2 Years