Flir IRW-2C Infrared Inspection Window, Aluminum, 2"

Representative image only.

Flir IRW-2C Infrared Inspection Window, Aluminum, 2"

Flir | Mfr: IRW-2C| Item: 3975490

Helps reduce the vast majority of dangerous arc flash occurrences

$381.61CAD / Each

$381.61CAD / Each
Product Description

Includes

window with hinged cover, gasket, and ring nut.

Key Features

  • Rugged frame handles both outdoor and indoor use
  • Decrease downtime and increase efficiency with easy installation
  • Easy flip-open hatch for faster inspections
  • Simple thumb screw releases the permanently hinged cover
  • Beaker features a 1000 mL capacity
  • Designed with graduation range from 100 to 1000 mL in 50 mL intervals
  • Features red colored markings
  • Limited Lifetime Warranty

More About this Item

Opening electrical cabinets to perform thermal and visual inspections of live components is dangerous work, exposing you to the risk of a hazardous arc flash incident. The FLIR IRW-series of thermal inspection windows help eliminate that danger. The window allows all brands of thermal imagers to capture both visible light and thermal images along with allowing LED and laser illumination to pass through the glass. A secure, permanently hinged cover opens easily with one hand, which means there’s nothing to remove, drop, or lose. The IRW-Series windows are comprehensively tested for safety and have a limited lifetime warranty.

Available for sale in the United States and Canada only

IP 67Limited Lifetime Warranty
Specifications
  • Max Temperature (° F)500
  • Max Temperature (° C)260
  • Camera IncludedNo
  • Height (in)3 23/64
  • Width (in)2 7/8
  • Length (in)1
  • DescriptionInfrared Inspection Window, Aluminum, 2"
  • WarrantyLimited Lifetime
Reviews
Q&A