Flir IRW-12PS Infrared Inspection Window, Powder-Coated Stainless Steel, 12"

Representative image only.

Flir IRW-12PS Infrared Inspection Window, Powder-Coated Stainless Steel, 12"

Flir | Mfr: IRW-12PS| Item: 3975807

Helps reduce the vast majority of dangerous arc flash occurrences

$1,339.31CAD / Each

$1,339.31CAD / Each
Product Description

Includes

window with hinged cover and gasket.

Key Features

  • Rugged frame handles both outdoor and indoor use
  • Decrease downtime and increase efficiency with easy installation
  • Easy flip-open hatch for faster inspections
  • Simple thumb screw releases the permanently hinged cover
  • Rectangular inspection window has a 12" width
  • Durable powder-coated stainless steel is ideal where mixed-metals are a concern and for outdoor applications
  • 1 year warranty

More About this Item

Opening electrical cabinets to perform thermal and visual inspections of live components is dangerous work, exposing you to the risk of a hazardous arc flash incident. The FLIR IRW-series of thermal inspection windows help eliminate that danger. The window allows all brands of thermal imagers to capture both visible light and thermal images along with allowing LED and laser illumination to pass through the glass. A secure, permanently hinged cover opens easily with one hand, which means there’s nothing to remove, drop, or lose. The IRW-Series windows are comprehensively tested for safety and have a limited lifetime warranty.

Available for sale in the United States and Canada only

IP 671 Year WarrantyLimited Lifetime Warranty
Specifications
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • Camera IncludedNo
  • Height (in)8 1/10
  • Width (in)12
  • DescriptionInfrared Inspection Window, Powder-Coated Stainless Steel, 12"
  • Warranty1 year
Reviews
Q&A