Extech UV510 Environmental-Series UV-A Light Meter

Representative image only.

Extech UV510 Environmental-Series UV-A Light Meter

Extech | Mfr: UV510| Item: 3000924

Measures UV-A light radiation from natural and artificial light sources

$300.57CAD / Each

$300.57CAD / Each
Product Description

Includes

Built-in light sensor with cover, wrist strap, tripod mount, and three AAA batteries.

Key Features

  • Record maximum and minimum values over time
  • Read measurements in low-lit areas on large, backlit LCD
  • Save battery life with auto-off setting
  • 1 year warranty

More About this Item

This compact UV light meter, with cosine correction sensor, measures irradiance from UV-A light sources up to 20 mW/cm2. It has a sensor wavelength range of 320 to 390 nm. Features include two selectable ranges, zero function, display Hold, and Min/Max readings.

1 Year WarrantyConformite Europeenne
Specifications
  • Resolution0.001 mW/cm²
  • Accuracy±4% FS
  • Display TypeBacklit dual LCD
  • SensorCosine correction
  • Height (in)5 1/2
  • Width (in)2 19/64
  • Depth (in)1
  • Height (cm)14.1
  • Width (cm)5.8
  • Depth (cm)2.5
  • PowerAAA Battery (3)
  • Min Measurement (mW/cm2)0
  • Max Measurement (mW/cm2)1.999
  • DescriptionUVA Light Meter
  • Warranty1 year
Reviews
Q&A