Extech UV505 Pocket UV-AB Light Meter

Representative image only.

Extech UV505 Pocket UV-AB Light Meter

Extech | Mfr: UV505| Item: 3572505

Compact and lightweight for quick and easy UV light measurement

$352.14CAD / Each

$352.14CAD / Each
Product Description

Includes

Protective sensor cap, carrying case, and two AAA batteries.

Key Features

  • 1 year warranty

More About this Item

Measure UV-A/B light from natural and artificial light sources in outdoor activities and indoor facilities—ideal for forensics, chromatography, electrophoresis, and electronics applications. UV sensor with cosine correction measures in the wavelength range of 290 to 390 nm. Zero point reset feature lets you easily zero using the protective cover.

1 Year WarrantyConformite Europeenne
Specifications
  • Resolution1 µW/cm²; 0.01 mW/cm²
  • Accuracy±4% full scale
  • FunctionZero point reset, hold, low battery
  • Display Type3-3/4-digit backlit LCD
  • SensorCosine correction
  • Height (in)2
  • Width (in)1 7/8
  • Depth (in)3/8
  • Depth (cm)0.9
  • Height (cm)5.2
  • Width (cm)4.8
  • PowerTwo AAA batteries
  • Min Measurement (µW/cm2)1
  • Max Measurement (µW/cm2)3999
  • Min Measurement (mW/cm2)0.01
  • Max Measurement (mW/cm2)39.99
  • DescriptionExtech UV505 Pocket UV-AB Light Meter
  • Warranty1 year
Reviews
Q&A