Extech ET11 GFCI Receptacle Tester with Pull Bar

Representative image only.

Extech ET11 GFCI Receptacle Tester with Pull Bar

Extech | Mfr: ET11| Item: 3975923

Safely test electrical sockets

$17.67CAD / Each

$17.67CAD / Each
Product Description

Key Features

  • Lights indicate circuit condition
  • Test for correct wiring of three-wire receptacles in 110 to 125 VAC, 50/60 Hz circuits
  • Detect five wiring faults: Open Ground, Open Neutral, Open Hot, Hot/Ground Reverse, Hot/Neutral Reverse
  • Confirms operation of the ground fault protective device
  • Ergonomic pull bar design for easy removal
  • Test code legend on both sides—easily verify both "ground bottom" and "ground top" style sockets
  • 3 year warranty

More About this Item

Quickly and safely test for faulty wiring in Type B electrical outlets with the receptacle testers. To use, simply plug the tester into a standard North American three-wire receptacle. Testers have indicator lights that light up instantly when faulty wiring is detected; lights also verify correctly wired receptacle. A test code light legend on the tester provides a quick and easy identification of potential hazards.

UL Listed.3 Year Warranty
Specifications
  • AC Max (Volts)120
  • Ac Voltage Accuracy±10%
  • Frequency Min (Hz)50
  • Frequency Max (Hz)60
  • DisplayIndicator lights
  • Interface3-wire with GCFI
  • MeasurementOpen ground, open neutral, open hot, hot & ground reversed, hot & neutral reversed,correct wiring
  • Safety RatingC-UL-US
  • Width (in)1.78
  • Length (in)4.26
  • Height (in)1.1
  • Width (cm)45
  • Length (cm)10.8
  • Height (cm)28
  • DescriptionGFCI Receptacle Tester with Pull Bar
  • Warranty3 years
Reviews
Q&A