Extech 451181 Printing Anemometer With Datalogging
Representative image only.
Extech 451181 Printing Anemometer With Datalogging
Extech | Mfr: 451181| Item: 1037999
$204.80CAD / Each
$204.80CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Measure, record, print and download air flow measuremetns to a PC
FEATURES
  • Simultaneously measures and displays air velocity, temperature, humidity, wet bulb, plus calculates CFM/CMM
  • Manually log measurements (99 points) or setup time interval for automatic logging (2400 points)
  • Datalogger date/time stamps and stores data for later recall, printing or downloading to a PC
  • Thermal printer prints air velocity, temperature, humidity, wet bulb, and CFM data with date and time stamp
  • Backlit display for operating in dimly lit areas
  • Auto power off
  • RS232 PC interface with Windows® 95/98/NT/2000/ME/XP compatible software to analyze data on PC
  • Complete with vane sensor with 4 ft (120cm) cable, software and cable, 4 AA batteries, and one paper roll
APPLICATIONS
  • Heating and air conditioning installation and repair
  • Airflow rate in air ducts
  • Air duct and ventilator analysis
  • Air quality monitor
  • Humidifier product testing
  • Mold remediation evaluation
1 Year Warranty
Specifications
  • BatteryFour AA
  • DescriptionPrinting Anemometer With Datalogging