Extech 382100 1200 A 3-Phase Power Analyzer/Datalogger

Representative image only.

Extech 382100 1200 A 3-Phase Power Analyzer/Datalogger

Extech | Mfr: 382100| Item: 2003652

Saves data on an SD card in Excel® format for easy transfer to a PC for analysis

$3,976.68CAD / Each

for an additional
$525.26CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$3,976.68CAD / Each

Buy It Together

Current Item

Extech 382100 1200 A 3-Phase Power Analyzer/Datalogger
Extech 382100 1200 A 3-Phase Power Analyzer/Datalogger
2003652

$3,976.68CAD / Each
+ =
Total Price 4501.94 ADD 2 ITEMS TO CART
Product Description

Includes

3 current clamps, 4 voltage leads with alligator clips, 8 AA batteries, SD memory card, Universal AC adaptor (100 to 240V), and case.

Key Features

  • Large dot-matrix, sun-readable, numerical, backlit LCD
  • Full system analysis with up to 35 parameters
  • V (phase-to-phase), V (phase-to-ground)
  • A (phase-to-ground)
  • KW / KVA / KVAR / PF (phase)
  • KW / KVA / KVAR / PF (system)
  • KWH / KVAH / KVARH / PFH (system)
  • Phase angle
  • High accuracy autoranging current clamps (0.2A to 1200A)
  • 600.0 VAC input with CAT III-600 V safety rating
  • Adjustable Current Transformer (CT) and Potential Transformer (PT) ratio for high power distribution systems
  • Log up to 30,000 reading on a removable SD memory card in Microsoft Excel® format
  • Wide sampling rate range (2 seconds to 2 hours)
  • Captured measurements can be imported directly into Excel via the SD memory card
  • Easy-to-use onscreen menu
  • Easy-to-grab rugged over molded housing
  • 1 year warranty
1 Year WarrantyConformite Europeenne
Specifications
  • Min AC Voltage (Voltage)0
  • Max AC Voltage (Voltage)600
  • AC Voltage Accuracy±(0.5% reading + 0.5V)
  • Min AC Current (amps)0
  • Max AC Current (amps)1200
  • AC Current Accuracy±(0.5% reading + 0.5 A)
  • Power Factor Accuracy±0.04
  • Min Frequency Range (Hz)45
  • Max Frequency Range (Hz)65
  • Frequency Accuracy±0.1 Hz
  • Active Power Range9.999 MW
  • Active Power Accuracy±(1.0% reading + 0.08 kW)
  • Apparent Power Range9.999 MVA
  • Apparent Power Accuracy±(1.0% reading + 0.08 kVA)
  • Reactive Power Range9.999 MVAR
  • Reactive Power Accuracy±(1.0% reading + 0.08 kVAR)
  • Power Factor Range0.00 to 1.00
  • Phase Angle-180 to 180°
  • Phase Accuracy±1°
  • Memory30,000 readings on SD card
  • Displaydot-matrix, backlit LCD
  • Battery Type8 AA (included)
  • Description1200 A 3-Phase Power Analyzer/Datalogger
  • Warranty1 Year
Reviews
Q&A