Expo Engineering TRX-55 Metal Band Drum Heater, 55 Gal, 200 to 400 °F, 3000 Watts, 240 V

Representative image only.

Expo Engineering TRX-55 Metal Band Drum Heater, 55 Gal, 200 to 400 °F, 3000 Watts, 240 V

Expo Engineering | Mfr: TRX-55-H/R-220V| Item: 0939954

Achieve better flow of viscous liquids

$1,137.46CAD / Each

$1,137.46CAD / Each
Product Description

Key Features

  • Three heat switch allows three wattage ratings per thermostat setting
  • Pilot light indicates when heating element is activated
  • Thermostat range from 200 to 4000 °F
  • 1 year warranty

More About this Item

These electric heaters combine adjustable temperature control with a variable power output selector. This output selector provides three wattage levels for a given thermostat setting and voltage rating. A pilot light indicates when the heating element is activated. Temperature detector is located on the outside of the pail or drum away from the active heating element.

Caution: Use heaters with metal pails or drums that contain nonflammable materials only. Heaters are only for indoor use in nonhazardous areas.

Fifty-five gallon drum heaters have a 6-ft, three-wire cord with high-wattage plug for 20-amp receptacle.

1 Year Warranty
Specifications
  • Drum MaterialMetal
  • Drum Size55 gallons
  • Max Temperature (° F)400
  • Max Temperature (° C)204
  • Min Temperature200;93
  • Plug TypeUS Standard
  • Power (VAC)240
  • Power (amps)12.5
  • Power (watts)3000
  • Width (in)4
  • Diameter (in)22.5 in
  • DescriptionDrum Heater, 55-Gallon, 200 to 400°F, 3000 Watts, 240 V
  • Warranty1 year
Reviews
Q&A