Expo Engineering TRX-5 Metal Band Pail Heater, 5 Gal, 200 to 400 °F, 1500 Watts, 120 V

Representative image only.

Expo Engineering TRX-5 Metal Band Pail Heater, 5 Gal, 200 to 400 °F, 1500 Watts, 120 V

Expo Engineering | Mfr: TRX-5-H/R| Item: 0939960

Achieve better flow of viscous liquids

$928.24CAD / Each

$928.24CAD / Each
Product Description

Includes

6-ft, three-wire cord with standard grounded plug.

Key Features

  • Three heat switch allows three wattage ratings per thermostat setting
  • Pilot light indicates when heating element is activated
  • Thermostat range from 200 to 400 °F
  • 1 year warranty

More About this Item

These electric heaters combine adjustable temperature control with a variable power output selector. This output selector provides three wattage levels for a given thermostat setting and voltage rating. A pilot light indicates when the heating element is activated. Temperature detector is located on the outside of the pail or drum away from the active heating element.

Caution: Use heaters with metal pails or drums that contain nonflammable materials only. Heaters are only for indoor use in nonhazardous areas.

1 Year Warranty
Specifications
  • Drum MaterialMetal
  • Drum Size5 gallons
  • Max Temperature (° F)400
  • Max Temperature (° C)204
  • Min Temperature200;93
  • Plug TypeUS Standard
  • Power (VAC)120
  • Power (amps)12.5
  • Power (watts)1500
  • Width (in)3
  • Diameter (in)11.25 in
  • DescriptionPail Heater, 5-Gallon, 200 to 400°F, 1500 Watts, 120 V
  • Warranty1 year
Reviews
Q&A