Erlab Pyramid Transportable Glove Enclosure

Representative image only.

Erlab Pyramid Transportable Glove Enclosure

Erlab | Mfr: 2200ANM| Item: 3366650

Protects you from hazardous, known, or potentially contaminated samples

$810.36CAD / Each

$810.36CAD / Each

Buy It Together

Current Item

Erlab Pyramid Transportable Glove Enclosure
Erlab Pyramid Transportable Glove Enclosure
3366650

$810.36CAD / Each
+
Cole-Parmer Economical Glove Box - Large
3478810

$6,615.52CAD / Each
=
Total Price 7425.88 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

1/8" ID coupler body valved fitting.

Key Features

  • Easily set up a controlled environment in the field or lab
  • Versatility to be used as test chamber
  • Folds into easy-to-carry size
  • Saves lab space
  • 1 year warranty

More About this Item

The Pyramid Transportable Glove Enclosure is design to protect you during the collection, transportation, investigation, and storage of samples or evidence. Final on-site testing processes can be safely performed as material remains sealed within an airtight chamber.

The portable light-weight glove enclosure with self-supporting pyramid assembles in seconds, collapses for easy storage, and can be cleaned with a mild cleanser for continued use. It features an 18 1/2” (47 cm) side opening for introduction of materials, a double-lined zipper to ensure an air-tight seal, and a port to exhaust/purge gas. It is constructed from high-quality transparent PVC and chemical resistant butyl rubber gloves with PVC sleeves. The slanted shape of the enclosure provides an ergonomic working position.

Each unit is individually tested to ensure its safety and comes with a unique serial number for full traceability.

The enclosure cannot be autoclaved.

1 Year Warranty
Specifications
  • Width of Equipment Sleeve (in)18.5
  • Width of Equipment Sleeve (cm)47
  • Chamber Width (in)28.5
  • Chamber Height (in)22
  • Chamber Depth (in)30
  • Exterior Width (in)8.6562
  • Exterior Height (in)30
  • Exterior Depth (in)22
  • Chamber Width (cm)72.39
  • Chamber Height (cm)55.88
  • Chamber Depth (cm)76.2
  • Exterior Width (cm)76.2
  • Exterior Height (cm)72.4
  • Exterior Depth (cm)55.9
  • DescriptionPyramid Transportable Glove Enclosure
  • Warranty1 year
Reviews
Q&A