Erlab 483 Resin Work Surface for Angled Sash Hoods, 49" W

Representative image only.

Erlab 483 Resin Work Surface for Angled Sash Hoods, 49" W

Erlab | Mfr: SMART481RT| Item: 3352061

$1,384.99CAD / Each

$1,384.99CAD / Each

Buy It Together

Current Item

Erlab 483 Resin Work Surface for Angled Sash Hoods, 49" W
Erlab 483 Resin Work Surface for Angled Sash Hoods, 49" W
3352061

$1,384.99CAD / Each
+ =
Total Price 8538.30 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from Replacement Filters, Work Surfaces, Carts, and Benches

Specifications
  • Hood Width (in)49
  • DescriptionWork Surface for Angled Sash Hoods, 49" W, resin
Reviews
Q&A