Erlab 481 Benchcap for Angled Sash Hoods, 49" W

Representative image only.

Erlab 481 Benchcap for Angled Sash Hoods, 49" W

Erlab | Mfr: SMART481BENCH| Item: 3352057

$1,952.24CAD / Each

$1,952.24CAD / Each

Buy It Together

Current Item

Erlab 481 Benchcap for Angled Sash Hoods, 49" W
Erlab 481 Benchcap for Angled Sash Hoods, 49" W
3352057

$1,952.24CAD / Each
+ =
Total Price 9105.55 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from Replacement Filters, Work Surfaces, Carts, and Benches

Specifications
  • Hood Width (in)49
  • DescriptionBase Stand for Angled Sash Hoods, 49" W
Reviews
Q&A