Erlab 392 Benchcap for Vertical Sash Hoods, 39" W

Representative image only.

Erlab 392 Benchcap for Vertical Sash Hoods, 39" W

Erlab | Mfr: SMART392BENCH| Item: 3352051

$1,908.04CAD / Each

$1,908.04CAD / Each

Buy It Together

Current Item

Erlab 392 Benchcap for Vertical Sash Hoods, 39" W
Erlab 392 Benchcap for Vertical Sash Hoods, 39" W
3352051

$1,908.04CAD / Each
+ =
Total Price 9061.35 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from Replacement Filters, Work Surfaces, Carts, and Benches

Specifications
  • Hood Width (in)39
  • DescriptionBase Stand for Vertical Sash Hoods, 39" W
Reviews
Q&A