Erlab 391 Glass Work Surface for Angled Sash Hoods, 39" W

Representative image only.

Erlab 391 Glass Work Surface for Angled Sash Hoods, 39" W

Erlab | Mfr: SMART391GL| Item: 3352045

$499.48CAD / Each

$499.48CAD / Each

Buy It Together

Current Item

Erlab 391 Glass Work Surface for Angled Sash Hoods, 39" W
Erlab 391 Glass Work Surface for Angled Sash Hoods, 39" W
3352045

$499.48CAD / Each
+ =
Total Price 7652.79 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

More About this Item

Choose from Replacement Filters, Work Surfaces, Carts, and Benches

Specifications
  • Hood Width (in)39
  • DescriptionWork Surface for Angled Sash Hoods, 39" W, glass
Reviews
Q&A