Erie Scientific Colorfrost® Plus Adhesion Microscope Slides, Blue; 144/Pk

Representative image only.

Erie Scientific Colorfrost® Plus Adhesion Microscope Slides, Blue; 144/Pk

Erie Scientific | Mfr: 5951PLUS-001| Item: 4851202

Tissue sections and cells electrostatically adhere to the glass without adhesives – eliminates tissue loss during staining

$274.42CAD / Pkg of 144

$274.42CAD / Pkg of 144
Product Description

Key Features

  • Color-coded for quick and simple identification
  • Marking surfaces ensure easy labeling
  • Slides measure 75 x 25 mm and are 1-mm thick
  • 1 year warranty

More About this Item

A permanent positive charge applied to the slide surface allows slides to hold tissue sections without the need for expensive or time-consuming preparatory steps in the laboratory

This treatment virtually eliminates tissue loss during staining and electrostatically attracts frozen tissue sections and cytology preparations, binding them to the slide.

1 Year Warranty
Specifications
  • Slide TypeBlue Colorfrost Plus
  • DescriptionBlue Colorfrost Plus Adhesion Slides, 144/pk
  • Warranty1 year
Reviews
Q&A