Environmental Express PD 2000 Benchtop Meter with Electrode Stand

Representative image only.

Environmental Express PD 2000 Benchtop Meter with Electrode Stand

Environmental Express | Item: 3541980

Developed using extensive feedback from laboratory staff, our new Environmental Express 2000 series meters are assembled in various configurations for measuring pH/mV, EC, Ion, DO, BOD parameters

$2,139.36CAD / Each

$2,139.36CAD / Each

Buy It Together

Current Item

Environmental Express PD 2000 Benchtop Meter with Electrode Stand
Environmental Express PD 2000 Benchtop Meter with Electrode Stand
3541980

$2,139.36CAD / Each
+ +
Environmental Express 2000 All-in-One pH Electrode
3541952

$282.89CAD / Each
+
Environmental Express pH 2000 Benchtop Meter Kit
3541954

$1,701.77CAD / Each
+ +
Environmental Express 2000 Replacement DO Sensor
3541975

$1,444.66CAD / Each
=
Total Price 7744.66 ADD 6 ITEMS TO CART
Product Description

Key Features

  • Various configurations available to meet the demands of measuring pH/mV, EC, Ion, PC, PD, DO, BOD parameters
  • Meter Kit formats include probes, 35419-54, -60, -65, -74, -78
  • Automatic record retention and accessibility for continuous compliance with regulatory requirements
  • 0.01, 0.001 resolution for routine or research applications where acute accuracy is critical
  • Large displays with a small footprint, ideal for labs with limited bench space
  • Automatic functions like buffer recognition and auto calibration for ease of use at any skill level

More About this Item

Environmental Express provides a diverse range of water quality analyzers and electrodes that are designed specifically for laboratory applications with compliance in mind. Our multi-parameter benchtops include functionality to meet the highly specific and/or broad ranging needs of wastewater, drinking water, food and beverage, chemical and pharmaceutical testing labs.

These benchtop meters offer a high-capacity internal memory that can capture and store 2000 data points with the time and date. They have a time-based auto data log with GLP compliant real time clocks. This data can be exported into CSV, Excel, or PDF formats via USB cable and use of complimentary software. A password access protection function secures the meter setup mode from unauthorized access. An adjustable shutoff time up to 30 minutes turns the meter off when idle.

Conformite Europeenne
Specifications
  • Meter TypepH/ORP/Ion/DO/BOD/Temp (°C/°F)
  • Display type5” Custom LCD with backlight and 320 segments
  • Connections2 x BNC, 2 x phono (ATC), DC sockets
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.1 / 0.01 / 0.001 pH
  • pH Accuracy± 0.003 pH
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 mV
  • mV Accuracy± 0.2 mV
  • Min ISE0.000 µg/L
  • Max ISE9999 g/L
  • ISE Resolution4 Significant digits
  • ISE Accuracy± 0.3% full scale or ± 0.2 mV, whichever is higher
  • Min Dissolved Oxygen0.0 mg/L or 0.0%
  • Max Dissolved Oxygen60.00 mg/L or 600.0%
  • Dissolved Oxygen Resolution0.01 mg/L, 0.1%
  • Dissolved Oxygen Accuracy+/- 0.1 mg/L , +/- 1.0%
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1 °C / °F
  • Temperature Accuracy± 0.5 °C / ± 0.9 °F
  • OutputPhono jack (USB/RS-232C)
  • Data Logging (points)2000
  • Power (VAC)100 to 240
  • Power (Hz)50-60 Hz
  • Length (cm)15.5
  • Width (cm)15
  • Height (cm)6.7
  • Width (in)5 29/32
  • Length (in)6 7/64
  • Height (in)2 41/64
  • Accessories IncludedElectrode stand
  • DescriptionPD 2000 Benchtop Meter with Electrode Stand
  • Warranty3 years
Reviews
Q&A