Environmental Express Oakton pH Electrode, 12x106 mm, Epoxy Body, Single Junction, Refill
Representative image only.
Environmental Express Oakton pH Electrode, 12x106 mm, Epoxy Body, Single Junction, Refill
Environmental Express | Item: 3580562
Affordable, high-accuracy pH electrodes for every application
$170.99CAD / Each
$170.99CAD / Each
Buy It Together
Current Item
Environmental Express Oakton pH Electrode, 12x106 mm, Epoxy Body, Single Junction, Refill
Environmental Express Oakton pH Electrode, 12x106 mm, Epoxy Body, Single Junction, Refill
3580562
$170.99CAD / Each
+ =
Total Price 397.67 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
3.3-ft (1-m) cable with a BNC connector.
Key Features
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
  • 1 year warranty
More About this Item
Environmental Express/ Oakton high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Electrode TypepH
  • Electrode Body MaterialEpoxy
  • Reference JunctionSingle
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)106
  • Cable Length (m)1
  • DescriptionpH Electrode, 12x106 mm, Epoxy Body, Single Junction, Refill