Environmental Express Oakton pH Electrode, 12x106 mm, Epoxy Body, Double Junction, Refill
Representative image only.
Environmental Express Oakton pH Electrode, 12x106 mm, Epoxy Body, Double Junction, Refill
Environmental Express | Item: 3580563
Affordable, high-accuracy pH electrodes for every application
$346.98CAD / Each
$346.98CAD / Each
Product Description
Includes
3.3-ft (1-m) cable with a BNC connector.
Key Features
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
  • 1 year warranty
More About this Item
Environmental Express/ Oakton high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Electrode TypepH
  • Electrode Body MaterialEpoxy
  • Reference JunctionDouble
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)106
  • Cable Length (m)1
  • DescriptionpH Electrode, 12x106 mm, Epoxy Body, Double Junction, Refill