Environmental Express Oakton Fast Response pH Electrode, 6x106 mm, Glass Body, Double Junction, Refill
Representative image only.
Environmental Express Oakton Fast Response pH Electrode, 6x106 mm, Glass Body, Double Junction, Refill
Environmental Express | Item: 3580568
Affordable, high-accuracy pH electrodes for every application
$370.19CAD / Each
$370.19CAD / Each
Product Description
Includes
3.3-ft (1-m) cable with a BNC connector.
Key Features
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
  • 1 year warranty
More About this Item
Environmental Express/ Oakton high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Electrode TypepH
  • Electrode Body MaterialGlass
  • Reference JunctionDouble
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)110
  • ConnectorBNC
  • Diameter (mm)6
  • Length (mm)106
  • Cable Length (m)1
  • DescriptionpH Electrode, 6x106 mm, Glass Body, Double Junction, Refill