Environmental Express Oakton Fast Response pH Electrode, 12x110 mm, Epoxy Body, Double Junction,Refillable
Representative image only.
Environmental Express Oakton Fast Response pH Electrode, 12x110 mm, Epoxy Body, Double Junction,Refillable
Environmental Express | Item: 3580569
Affordable, high-accuracy pH electrodes for every application
$262.26CAD / Each
$262.26CAD / Each
Buy It Together
Current Item
Environmental Express Oakton Fast Response pH Electrode, 12x110 mm, Epoxy Body, Double Junction,Refillable
Environmental Express Oakton Fast Response pH Electrode, 12x110 mm, Epoxy Body, Double Junction,Refillable
3580569
$262.26CAD / Each
+ =
Total Price 488.94 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
3.3-ft (1-m) cable with a BNC connector.
Key Features
  • The ideal 5-point calibration pH electrode
  • Delivers superior reproducibility in aqueous samples
  • 1 year warranty
More About this Item
Environmental Express/ Oakton high-performance electrodes are available in a full range of materials, junctions, and sizes to suit your application requirements. New formulation provides additional stability and superior reproducibility.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Electrode TypepH
  • Electrode Body MaterialEpoxy
  • Reference JunctionDouble
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)106
  • Cable Length (m)1
  • DescriptionpH Electrode, 12x110 mm, Epoxy Body, Double Junction, Sealed