Environmental Express Oakton Buffer Capsules, pH 10; 10/Pk
Representative image only.
Environmental Express Oakton Buffer Capsules, pH 10; 10/Pk
Fresh pH calibration solution anytime and anywhere
$43.17CAD / Pkg of 10
$43.17CAD / Pkg of 10
Buy It Together
Current Item
Environmental Express Oakton Buffer Capsules, pH 10; 10/Pk
Environmental Express Oakton Buffer Capsules, pH 10; 10/Pk
3585016
$43.17CAD / Pkg of 10
+
Environmental Express Electrode Storage Solution; 500 mL
0065304
$40.22CAD / Pint of 1
=
Total Price 83.39 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Ten blue-colored capsules.
Key Features
  • Each capsule mixes with 100-mL of deionized water to create a clear, 10.00 pH solution
  • 3 month warranty
More About this Item
Buffer salts inside capsules require 100 mL of deionized or distilled water to produce a clear pH calibration solution accurate to ±0.02 pH (at 25°C) with traceability to NIST standards. Capsules are easier to store and transport than prepared aqueous buffer solutions. Capsules are color-coded for easy identification if mixed.
Store buffer solution in an airtight container at room temperature for three weeks. Unused capsules have a five-year shelf life. Each vial contains 10 capsules.
3 Month Warranty
Specifications
  • Buffer Value10
  • Volume (mL)100
  • ColorClear
  • DescriptionBuffer Capsules, pH 10; 10/Pk